Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPAVLGFEG SANKIGVGVV RDGKVLANPR RTYVTPPGTG FLPGDTARHH RAVILDLLQE ALTESGLTSQ DIDCIAYTKG PGMGAPLVSV AVVARTVAQL WNKPLVGVNH CIGHIEMGRL ITGATSPTVL YVSGGNTQVI AYSEHRYRIF GETIDIAVGN CLDRFARVLK ISNDPSPGYN IEQMAKRGKK LVELPYTVKG MDVSFSGILS FIEDVAHRML ATGECTPEDL CFSLQETVFA MLVEITERAM AHCGSQEALI VGGVGCNVRL QEMMATMCQE RGARLFATDE RFCIDNGAMI AQAGWEMFRA GHRTPLSDSG VTQRYRTDEV EVTWRD (1-335 aa encoded by B C032310)
Osgep (O-sialoglycoprotein endopeptidase), also known as GCPL1, is a 335 amino acid protein that is a member of the peptidase M22 family. Osgep specifically cleaves the 31-Arg-l-Asp-32 bond in glycophorin A, but it does not cleave desialylated glycoproteins, unglycosylated proteins or glycoproteins that are only N-glycosylated. Though ubiquitously expressed at low levels, highest levels of Osgep are found in liver, skeletal muscle and kidney. OSGEPL1, also known as Qri7, is a 414 amino acid protein that belongs to the KAE1/YgjD family and exists as three alternatively spliced isoforms. In tRNAs that have codons beginning with adenine, OSGEPL1 is required for the formation of a threonylcarbamoyl group on adenosine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hOSGEP; mOsgep; N6-L-threonylcarbamoyladenine synthase; O-sialoglycoprotease; O-sialoglycoprotein endopeptidase; O-sialoglycoprotein endopeptidase {ECO:0000255|HAMAP-Rule:MF_03180}; OSGEP; pasteurella haemolytica metalloprotease homolog with glycoprotein substrates/gpc-like protein 1; probable O-sialoglycoprotein endopeptidase; probable tRNA N6-adenosine threonylcarbamoyltransferase; probable tRNA N6-adenosine threonylcarbamoyltransferase {ECO:0000255|HAMAP-Rule:MF_03180}; probable tRNA threonylcarbamoyladenosine biosynthesis protein OSGEP; Prsmg1/Gcpl1; t(6)A synthase; t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEP; t(6)A37 threonylcarbamoyladenosine biosynthesis protein Osgep {ECO:0000255|HAMAP-Rule:MF_03180}; tRNA N6-adenosine threonylcarbamoyltransferase; tRNA threonylcarbamoyladenosine biosynthesis protein OSGEP; tRNA threonylcarbamoyladenosine biosynthesis protein Osgep {ECO:0000255|HAMAP-Rule:MF_03180}
Gene Aliases: 1500019L24Rik; GCPL-1; GCPL1; KAE1; OSGEP; OSGEP1; PRSMG1
UniProt ID: (Human) Q9NPF4, (Rat) Q9WVS2, (Mouse) Q8BWU5
Entrez Gene ID: (Human) 55644, (Rat) 290028, (Mouse) 66246
Molecular Function:
RNA processing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support