Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GLPTVPSDHL PNLYGFSALH AVHLHQWTLG YPAMHLPRSS FSKVPGTVSS LVDARFQLPA FPWFPHVIQP KPEITAGGSV
OSR1 is a transcription factor that plays a role in the regulation of embryonic heart and urogenital development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: odd-skipped homolog; odd-skipped related 1; Protein odd-skipped-related 1
Gene Aliases: ODD; OSR1
UniProt ID: (Human) Q8TAX0
Entrez Gene ID: (Human) 130497
Molecular Function: zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support