Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VQLKGRDLL TLKNFTGEEI KYMLWLSADL KFRIKQKGEY LPLLQGKSLG MIFEKRSTRT RLSTETGFAL LGGHPCFLTT QDIHLGVNES LTDTARVLSS MADAVLARVY KQSDLDTLAK EASIPIINGL SDLYHPIQIL ADYLTLQEHY SSLKGLTLSW IGDGNNILHS IMMSAAKFGM HLQAATPKGY EPDASVTKLA EQYAKENGTK LLLTNDPLEA AHGGNVLITD TWISMGQEEE KKKRLQAFQG YQVTMKTAKV AASDWTFLHC LPRKPEEVDD EVFYSPRSLV FPEAENRKWT IMAVMVSLLT DYSPQLQKPK F (35-354 aa encoded by BC074745)
This nuclear gene encodes a mitochondrial matrix enzyme. Missense, nonsense, and frameshift mutations in this enzyme lead to ornithine transcarbamylase deficiency, which causes hyperammonemia. Since the gene for this enzyme maps close to that for Duchenne muscular dystrophy, it may play a role in that disease also.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ornithine carbamoyltransferase, mitochondrial; ornithine transcarbamylase; Ornithine transcarbamylase, mitochondrial; OTCase; sparse fur
Gene Aliases: AI265390; OCTD; OTC; Sf; spf
UniProt ID: (Human) P00480, (Mouse) P11725, (Rat) P00481
Entrez Gene ID: (Human) 5009, (Mouse) 18416, (Rat) 25611
Molecular Function: transferase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support