Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the homeodomain (HD) family. HD family proteins are helix-turn-helix transcription factors that play key roles in the specification of cell fates. This protein may function during brain development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein orthopedia; orthopedia homolog
Gene Aliases: OTP
UniProt ID: (Human) Q5XKR4, (Mouse) O09113
Entrez Gene ID: (Human) 23440, (Mouse) 18420, (Rat) 294640
Molecular Function:
helix-turn-helix transcription factor
homeodomain transcription factor
DNA-binding transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support