Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
May play an important role in the cellular response to tissue hypoxia. May be either a GDP-fucose transporter that competes with SLC35C1 for GDP-fucose, or a factor that otherwise enhances the fucosylation of Notch and is required for optimal Notch signaling in mammalian cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ovarian cancer overexpressed gene 1 protein; Ovarian cancer-overexpressed gene 1 protein; solute carrier family 35 (GDP-fucose transporter), member C2; Solute carrier family 35 member C2
Gene Aliases: BA394O2.1; C20orf5; C85957; CGI-15; D2Wsu58e; OVCOV1; SLC35C2
UniProt ID: (Human) Q9NQQ7, (Mouse) Q8VCX2
Entrez Gene ID: (Human) 51006, (Mouse) 228875
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support