Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 10
OX40 is a protein receptor found on the surface of T cells, a type of white blood cell that plays a crucial role in the immune system. When activated, OX40 promotes the proliferation and survival of T cells, leading to a stronger immune response against cancer cells and infectious agents. OX40 agonists, which are drugs that activate OX40, have shown promising results in preclinical and clinical studies as a potential immunotherapy for cancer. They can enhance the efficacy of other cancer treatments such as chemotherapy and checkpoint inhibitors, and may also have applications in autoimmune diseases and allergies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; lymphoid activation antigene ACT35; MRC OX40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; Tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; tumor necrosis factor (ligand) superfamily member 4; Tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4
Gene Aliases: ACT35; CD134; IMD16; Ly-70; OX40; TNFRSF4; Txgp1; TXGP1L
UniProt ID: (Human) P43489, (Rat) P15725, (Mouse) P47741
Entrez Gene ID: (Human) 7293, (Rat) 25572, (Mouse) 22163
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support