Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MYWLSSNLF SLVQVSCLRI PAVRTVLKIP QRVVHDLDKL PPREGFLESF KKGWKNAEMT RQLREREQRM RNQLELAARG PLRQTFTHNP LLQPGKDNPP NIPSSSSKPK SKYPWHDTLG (319-438 aa encoded by BC001669)
OXA1L is required for the insertion of integral membrane proteins into the mitochondrial inner membrane. It is essential for the activity and assembly of cytochrome oxidase and is required for the correct biogenesis of ATP synthase and complex I in mitochondria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hsa; Mitochondrial inner membrane protein OXA1L; OXA1-like protein; OXA1Hs; oxidase (cytochrome c) assembly 1-like; Oxidase assembly 1-like protein
Gene Aliases: OXA1; OXA1L
UniProt ID: (Human) Q15070
Entrez Gene ID: (Human) 5018
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support