Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YGTLVQEGG APIRYTPDGH LALMSQPREV REFNGDHFLL ERAIRADFAL VKGWKADRAG NVVFRRSARN FNVPMCKAAD VTAVEVEEIV EVGAFPPEDI HVPNIYVDRV IKGQKYEKRI ERLTILKEED GDAGKEEDAR TRIIRRAALE FEDGMYANLG IGIPLLASNF ISPSMTVHLH SENGILGLGP FPTEDEVDAD LINAGKQTVT VLPGGCFFAS DDSFAMIRGG HIQLTMLGAM QVSKYGDLAN WMIPGKKVKG MGGAMDLVSS QKTRVVVTMQ HCTKDNTPKI MEKCTMPLTG KRCVDRIITE KAVFDVHRKK ELTLRELWEG LTVDDIKKST GCAFAVSPNL RPMQ (161-517 aa encoded by B C030015)
Key enzyme for ketone body catabolism. Transfers the CoA moiety from succinate to acetoacetate. Formation of the enzyme-CoA intermediate proceeds via an unstable anhydride species formed between the carboxylate groups of the enzyme and substrate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-oxoacid CoA transferase 2; 3-oxoacid CoA-transferase 2A; 3-oxoacid-CoA transferase 2A; haploid germ cell specific succinyl CoA transferase; SCOT-t; SCOT-t1; Succinyl-CoA:3-ketoacid coenzyme A transferase 2, mitochondrial; Succinyl-CoA:3-ketoacid coenzyme A transferase 2A, mitochondrial; succinyl-CoA:3-ketoacid-coenzyme A transferase 2, mitochondrial; succinyl-CoA:3-ketoacid-coenzyme A transferase 2A, mitochondrial; succinyl-CoA:3-oxoacid CoA-transferase; testis-specific succinyl CoA:3-oxoacid CoA-transferase; testis-specific succinyl CoA:3-oxoacid CoA-transferase 1; Testis-specific succinyl-CoA:3-oxoacid CoA-transferase; Testis-specific succinyl-CoA:3-oxoacid CoA-transferase 1
Gene Aliases: FKSG25; Oxct; OXCT2; Oxct2a; Scot; Scot-t1; SCOTT
UniProt ID: (Human) Q9BYC2, (Rat) Q5XIJ9, (Mouse) Q9JJN4
Entrez Gene ID: (Human) 64064, (Rat) 366463, (Mouse) 64059
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support