Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAALRLLASV LGRGVPAGGS GLALSQGCAR CFATSPRLRA KFYADPVEMV KDISDGATVM IGGFGLCGIP ENLIAALLRT RVKDLQVVSS NVGVEDFGLG LLLAARQVRR IVCSYVGENT LCESQYLAGE LELELTPQGT LAERIRAGGA GVPAFYTPTG YGTLVQEGGA PIRYTPDGHL ALMSQPREVR EFNGDHFLLE RAIRADFALV KGWKADRAGN VVFRRSARNF NVPMCKAADV TAVEVEEIVE VGAFPPEDIH VPNIYVDRVI KGQKYEKRIE RLTILKEEDG DAGKEEDART RIIRRAALEF EDGMYANLGI GIPLLASNFI SPSMTVHLHS ENGILGLGPF PTEDEVDADL INAGKQTVTV LPGGCFFASD DSFAMIRGGH IQLTMLGAMQ VSKYGDLANW MIPGKKVKGM GGAMDLVSSQ KTRVVVTMQH CTKDNTPKIM EKCTMPLTGK RCVDRIITEK AVFDVHRKKE LTLRELWEGL TVDDIKKSTG CAFAVSPNLR PMQ
Key enzyme for ketone body catabolism. Transfers the CoA moiety from succinate to acetoacetate. Formation of the enzyme-CoA intermediate proceeds via an unstable anhydride species formed between the carboxylate groups of the enzyme and substrate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-oxoacid CoA transferase 2; 3-oxoacid CoA-transferase 2A; 3-oxoacid-CoA transferase 2A; SCOT-t; Succinyl-CoA:3-ketoacid coenzyme A transferase 2, mitochondrial; succinyl-CoA:3-ketoacid-coenzyme A transferase 2, mitochondrial; testis-specific succinyl CoA:3-oxoacid CoA-transferase; Testis-specific succinyl-CoA:3-oxoacid CoA-transferase
Gene Aliases: FKSG25; OXCT2; SCOTT
UniProt ID: (Human) Q9BYC2
Entrez Gene ID: (Human) 64064
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support