Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: U87-MG.
Predicted reactivity: Mouse (83%), Rat (85%), Rhesus Monkey (96%), Bovine (86%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
OXNAD1 is a protein coding gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: oxidoreductase NAD-binding domain containing 1; Oxidoreductase NAD-binding domain-containing protein 1
Gene Aliases: OXNAD1
UniProt ID: (Human) Q96HP4
Entrez Gene ID: (Human) 92106
Molecular Function:
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support