Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to OCT6.
POU3F1 is a member of the POU domain family of proteins, and regulates events during neurogenesis and myelination.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 6-Oct; Oct-6; octamer binding factor oct6; Octamer-binding protein 6; Octamer-binding transcription factor 6; octoamer transcription factor-6; octomer-6; OTF-6; POU domain transcription factor SCIP; POU domain, class 3, transcription factor 1; suppressed cAMP-inducible POU; Tst-1
Gene Aliases: Oct-6; OCT6; Otf-6; OTF6; POU3F1; SCIP; Test1; Testes-1; Tst-1; Tst1
UniProt ID: (Human) Q03052, (Mouse) P21952, (Rat) P20267
Entrez Gene ID: (Human) 5453, (Mouse) 18991, (Rat) 192110
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support