Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Endotoxin level < 0.01EU/µg by LAL method
Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Olaratumab (IMC-3G3) is a fully human IgG1 monoclonal antibody with antitumor activity that selectively binds the external domain of human platelet-derived growth factor receptor (PDGFR)-alpha with high affinity and blocks ligand binding. It is composed of two heavy chain molecule fragments and 2 light chain fragments. Studies show that the treatment of olaratumab in combination with doxorubicin resulted in significant reduction of cancer cell proliferation and tumor growth. Olaratumab was granted accelerated approval (as Lartruvo) as initial therapy to treat adults with certain types of soft tissue sarcoma (STS) in October, 2016.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3G3; IMC-3G3; LY3012207
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support