Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total Olfactory receptor 51I1.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GA_x6K02T2PBJ9-6756759-6755815; HOR5'Beta11; Odorant receptor HOR5'beta11; olfactory receptor; Olfactory receptor 51I1; olfactory receptor 640; olfactory receptor MOR13-4; Olfactory receptor OR11-39; olfactory receptor, family 51, subfamily I, member 1
Gene Aliases: MOR13-4; OR51I1
UniProt ID: (Human) Q9H343
Entrez Gene ID: (Human) 390063, (Mouse) 258819
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support