Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC103389; ODC; Ornithine decarboxylase; Ornitine decarboxylase
Gene Aliases: ODC; ODC1
UniProt ID: (Human) P11926, (Rat) P09057, (Mouse) P00860
Entrez Gene ID: (Human) 4953, (Rat) 24609, (Mouse) 18263
Molecular Function:
decarboxylase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support