Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 9
Antibody detects endogenous levels of total OSR2.
OSR2 is a mammalian homolog of the Drosophila odd-skipped family of transcription factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: odd-skipped related 2; Protein odd-skipped-related 2
Gene Aliases: 5430409I15Rik; OSR2; Osr2A; Osr2B
UniProt ID: (Human) Q8N2R0, (Mouse) Q91ZD1, (Rat) Q6AY34
Entrez Gene ID: (Human) 116039, (Mouse) 107587, (Rat) 315039
Molecular Function:
zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support