Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 50 µL of distilled water (2 mg/mL final concentration) then dilute further for applications in PBS or media that is identical to that used in the assay system.
Osteocalcin belongs to the osteocalcin/matrix Gla-protein family and constitutes 1-2% of the total bone protein. It binds strongly to apatite and calcium. Gamma-carboxyglutamate residues are formed by vitamin K dependent carboxylation. These residues are essential for the binding of calcium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BGP; bone gamma-carboxyglutamate (gla) protein; bone gamma-carboxyglutamate (gla) protein (osteocalcin); bone gamma-carboxyglutamate protein 2; bone gamma-carboxyglutamate protein osteocalcin; bone gamma-carboxyglutamate; BGLAP; bone gamma-carboxyglutamic acid (Gla) protein (osteocalcin); Bone Gla protein; bone gla protein BGP; bone Gla-protein; gamma-carbooxyglutamate (gla) protein; Gamma-carboxyglutamic acid-containing protein; Gla; Osteocalcin; PMF1; RP11-54H19.5
Gene Aliases: BGLAP; Bglap2; BGP; Bgpr; Bgpra; GLA; OC; OCN
UniProt ID: (Human) P02818, (Rat) P04640
Entrez Gene ID: (Dog) 403762, (Chicken) 396348, (Sheep) 677670, (Human) 632, (Rabbit) 100340503, (Pig) 397530, (Goat) 100861188, (Rat) 25295
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support