Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute at 0.5 mg/mL in sterile PBS.
Hormone that acts as a regulator of dendritic growth in the developing cerebral cortex in response to sensory experience. Induced in the brain following membrane depolarization and inhibits dendritic branching in neurons of the developing cortex
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Musclin; Osteocrin
Gene Aliases: MUSCLIN; Ostc; OSTN
UniProt ID: (Human) P61366, (Mouse) P61364
Entrez Gene ID: (Human) 344901, (Mouse) 239790
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support