Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in rat and mouse.
Specificity of this antibody: Otoferlin.
Otoferlin is a protein that in humans is encoded by the OTOF gene. Mutations in this gene are a cause of neurosensory nonsyndromic recessive deafness, DFNB9. The short form of the encoded protein has three C2 domains, a single carboxy-terminal transmembrane domain found also in the C. elegans spermatogenesis factor FER-1 and human dysferlin, while the long form has six C2 domains. The homology suggests that this protein may be involved in vesicle membrane fusion. Several transcript variants encoding multipleisoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fer-1-like family member 2; Fer-1-like protein 2; Otoferlin; Protein pachanga
Gene Aliases: AUNB1; DFNB6; DFNB9; FER1L2; NSRD9; OTOF
UniProt ID: (Human) Q9HC10, (Rat) Q9ERC5, (Mouse) Q9ESF1
Entrez Gene ID: (Human) 9381, (Rat) 84573, (Mouse) 83762
Molecular Function: membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support