Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive control: Ovotransferrin
Transferrins (Tf) are an approximately 80kDa blood plasma glycoprotein synthesized by the liver. They are iron binding transport proteins which can bind two Fe(3+) ions in association with the binding of an anion, usually bicarbonate. It is responsible for the transport of iron from sites of absorption and heme degradation to those of storage and utilization. Transferrin is the primary blood iron transport protein and under normal conditions; approximately one-third of total blood transferrin contains bound iron. Measurement of blood transferrin levels can be used as an indicator for blood iron-carrying capacity and abnormalities of iron metabolism such as anaemia, iron overload and haemochromatosis.Serum transferrin may also have a further role in stimulating cell proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Allergen Gal d III; Beta-1 metal-binding globulin; Conalbumin; DKFZp781D0156; epididymis secretory sperm binding protein Li 71p; lactotransferrin; Ovotransferrin; ovotransferrin BB type; ovotransferrin BC type; ovotransferrin CC type; Serotransferrin; Serum transferrin; Siderophilin; transferin; Transferrin
Gene Aliases: conalbumin; HEL-S-71p; LTF; PRO1400; PRO1557; PRO2086; TF; TFEW; TFQTL1
UniProt ID: (Human) P02787, (Chicken) P02789
Entrez Gene ID: (Human) 7018, (Chicken) 396241
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support