Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TDSVVSSVT TKVKGVAVTN TSKLGFRIWD VADYVIPAQE ENSLFVMTNV ILTMNQTQGL CPEIPDATTV CKSDASCTAG SAGTHSNGVS TGRCVAFNGS VKTCEVAAWC PVEDDTHVPQ PAFLKAAENF TLLVKNNIWY PKFNFSKRNI LPNITTTYLK SCIYDAKTDP FCPIFRLGKI VENAGHGFQD MAVEGGIMGI QVNWDCNLDR AASLCLPRYS FRRLDTRDVE HNVSPGYNFR FAKYYRDLAG NEQRTLIKAY GIRFDIIVFG KAGKFDIIPT MINIGS (57-341 aa encoded by BC033826)
The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel with high calcium permeability. The main pharmacological distinction between the members of the purinoceptor family is the relative sensitivity to the antagonists suramin and PPADS. The product of this gene has the lowest sensitivity for these antagonists. Multiple alternatively spliced transcript variants have been identified for this gene although their full-length natures have not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP receptor; ATP-gated cation channel protein; ionotropic purinergic receptor; P2X purinoceptor 4; P2X receptor, subunit 4; P2X4; Purinergic receptor; purinergic receptor P2X, ligand gated ion channel, 4; purinergic receptor P2X, ligand-gated ion channel, 4; purinergic receptor P2X4; purinergic receptor P2x4 subunit variant e; purinoceptor P2X4
Gene Aliases: AI504491; AW555605; D5Ertd444e; P2RX4; P2X4; P2X4R
UniProt ID: (Human) Q99571, (Rat) P51577
Entrez Gene ID: (Pig) 100157723, (Human) 5025, (Mouse) 18438, (Rat) 29659
Molecular Function:
ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support