Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C, protected from the light, for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 X g 5 min).
P2Y1 is a Purinergic Receptor that causes a change in platelet shape and potentially leads to platelet aggregation by binding ADP, which results in the mobilization of intracellular calcium ions via activation of phospholipase C. This receptor also mediates the release of the relaxing factor nitric oxide and may be involved in the modulation of neuronal signal transmission. P2Y1 has been reported to be expressed in tissues throughout the body, particularly brain. ESTs have been isolated from human embryo, lung, and placenta libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP receptor; ATP receptor; P2 purinoceptor subtype Y1; P2 purinoreceptor subclass 2Y; P2Y ATP receptor 1; P2Y purinoceptor 1; p2y-1-r; P2Y1; P2Y1 receptor; p2y1r; platelet ADP receptor; Purinergic receptor; purinergic receptor P2Y, G-protein coupled, 1; Purinoceptor p2y1; Purinoceptor p2yr1
Gene Aliases: P2RY1; P2y; P2Y1
UniProt ID: (Human) P47900, (Rat) P49651, (Mouse) P49650
Entrez Gene ID: (Human) 5028, (Rat) 25265, (Mouse) 18441
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support