Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: SK-Br-3, A549, human liver tissue, human colon tissue, human kidney tissue.
P2Y6 belongs to the family of G protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is responsive to UDP, partially responsive to UTP and ADP, and not responsive to ATP. P2Y6 expression has been documented in various blood cells, bone marrow, brain, colon, heart, kidney, ovary, placenta, spleen and thymus. ESTs have been isolated from cervix, embryo, eye, kidney, liver/spleen, placenta, and vessel libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G-coupled nucleotide receptor; MGC15335; P2 purinoceptor; P2Y purinoceptor 6; p2y3 and chick; p2y3 receptor; P2Y6; P2Y6 receptor; Purinoceptor p2y6; pyrimidinergic receptor P2Y, G-protein coupled, 6
Gene Aliases: P2RY6; P2Y6; PP2891
UniProt ID: (Human) Q15077
Entrez Gene ID: (Human) 5031
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support