Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Amino acid sequence of the immunogen is proprietary. This antibody has been tested in indirect ELISA.
P2Y8 belongs to the family of G-protein coupled receptors, that are preferentially activated by adenosine and uridine nucleotides. The gene codifying for this protein is moderately expressed in undifferentiated HL60 cells, and is located at both chromosomes X and Y.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G-protein coupled purinergic receptor P2Y8; MGC50878; P2Y purinoceptor 8; P2Y8; purinergic receptor P2Y, G-protein coupled, 8; RP11-261P4.4
Gene Aliases: P2RY8; P2Y8
UniProt ID: (Human) Q86VZ1
Entrez Gene ID: (Human) 286530
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support