Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HTVLWMEQV LKQLDAGEEA TTTKSQVLDY LSYAVFQLGD LHRALELTRR LLSLDPSHER AGGNLRYFEQ LLEEEREKTL TNQTEAELAT PEGIYERPVD YLPERDVYES LCRGEGVKLT PRRQKRLFCR YHHGNRAPQL LIAPFKEEDE WDSPHIVRYY DVMSDEEIER IKEIAKPKLA RATVRDPKTG VLTVASYRVS KSSWLEEDDD PVVARVNRRM QHITGLTVKT AELLQVANYG VGGQYEPHFD FSRRPFDSGL KTEGNRLATF LNYMSDVEAG GATVFPDLGA AIWPKKGTAV FWYNLLRSGE GDYRTRHAAC PVLVGCKWVS NKWFHERGQE FLRPCGSTEV D (184-533 aa encoded by BC035813)
This gene encodes a component of prolyl 4-hydroxylase, a key enzyme in collagen synthesis composed of two identical alpha subunits and two beta subunits. The encoded protein is one of several different types of alpha subunits and provides the major part of the catalytic site of the active enzyme. In collagen and related proteins, prolyl 4-hydroxylase catalyzes the formation of 4-hydroxyproline that is essential to the proper three-dimensional folding of newly synthesized procollagen chains. Alternatively spliced transcript variants encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4-PH alpha 2; 4-PH alpha-2; 4PHalpha2; C-P4Halpha(II); collagen prolyl 4-hydroxylase alpha(II); procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha II polypeptide; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide II; Procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-2; Prolyl 4-hydroxylase alpha IIa subunit; Prolyl 4-hydroxylase subunit alpha-2; prolyl 4-hydroxylase, alpha polypeptide II
Gene Aliases: AA407196; C76437; P4HA2; P4hl; UNQ290/PRO330
UniProt ID: (Human) O15460, (Mouse) Q60716
Entrez Gene ID: (Human) 8974, (Mouse) 18452, (Rat) 360526
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support