Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: TRFYDKVLSL HEDSTTPVAN PLLAFTLIKR LQSDWRNVVH SLEASENIRA LKDGYEKVEQ DLPAFEDLEG AARALMRLQD VYMLNVKGLA RGVFQRVTGS
Catalyzes the post-translational formation of 4- hydroxyproline in -Xaa-Pro-Gly- sequences in collagens and other proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4-PH alpha-3; C-P4H alpha III; collagen prolyl 4-hydroxylase alpha(III); procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide III; Procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-3; Prolyl 4-hydroxylase subunit alpha-3; prolyl 4-hydroxylase, alpha polypeptide III
Gene Aliases: P4HA3; UNQ711/PRO1374
UniProt ID: (Human) Q7Z4N8
Entrez Gene ID: (Human) 283208
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support