Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Catalyzes the post-translational formation of 4- hydroxyproline in -Xaa-Pro-Gly- sequences in collagens and other proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4-PH alpha-3; C-P4H alpha III; collagen prolyl 4-hydroxylase alpha 3; collagen prolyl 4-hydroxylase alpha III subunit; collagen prolyl 4-hydroxylase alpha(III); procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide III; Procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-3; Prolyl 4-hydroxylase subunit alpha-3; prolyl 4-hydroxylase, alpha polypeptide III
Gene Aliases: D930031A02Rik; P4HA3; UNQ711/PRO1374
UniProt ID: (Human) Q7Z4N8, (Rat) Q6W3E9
Entrez Gene ID: (Human) 283208, (Mouse) 320452, (Rat) 361612
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support