Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
The poly(A)-binding protein (PABP), which is found complexed to the 3-prime poly(A) tail of eukaryotic mRNA, is required for poly(A) shortening and translation initiation. In humans, the PABPs comprise a small nuclear isoform and a conserved gene family that displays at least 3 functional proteins: PABP1 (PABPC1), inducible PABP (iPABP, or PABPC4), and PABP3 (PABPC3). In addition, there are at least 4 pseudogenes, PABPCP1 to PABPCP4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PABP 1; PABP-1; poly(A) binding protein, cytoplasmic 1; poly(A) binding protein, cytoplasmic 2; poly(A)-binding protein 1; Polyadenylate-binding protein 1
Gene Aliases: PAB1; PABP; PABP1; PABPC1; PABPC2; PABPL1
UniProt ID: (Human) P11940, (Rat) Q9EPH8
Entrez Gene ID: (Human) 26986, (Rat) 171350
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support