Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SARSHKEPPS PADVPEKTRS LGGRQPSDSV SDTVALGVPG PREHPGQPED SPEAEASTLD VFTERLPPSG RITKTESLVI PSTRSEGKQA GRRGRSTSLK ERQAARPQNE RANSLDNERC PDARSQLQIP RKTVYDQLNH ILISDDQLPE NIILVNTSDW Q
PACS2 is the multifunctional sorting protein that controls the endoplasmic reticulum (ER)-mitochondria communication, including the apposition of mitochondria with the ER and ER homeostasis. In addition, in response to apoptic inducer, PACS2 translocates BIB to mitochondria, which initiates a sequence of events including the formation of mitochondrial truncated BID, the release of cytochrome c, the activation of caspase-3 thereby causing cell death.PACS2 may also be involved in ion channel trafficking, directing acidic cluster-containing ion channels to distinct subcellular compartments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ25488; PACS-2; PACS1-like protein; Phosphofurin acidic cluster sorting protein 2
Gene Aliases: KIAA0602; PACS-2; PACS1L; PACS2
UniProt ID: (Human) O60342
Entrez Gene ID: (Human) 23241
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support