Product References
Inhibition of protein arginine deiminase II suppresses retinoblastoma in orthotopic transplantation in mice.
Oncology reports
Kim S,Song YK,Cho CS,Kim HJ,Fang S,Jo DH,Kim H
Sat Jul 01 00:00:00 EDT 2023
Inhibition of protein arginine deiminase II suppresses retinoblastoma in orthotopic transplantation in mice.
Oncology reports
Kim S,Song YK,Cho CS,Kim HJ,Fang S,Jo DH,Kim H
Sat Jul 01 00:00:00 EDT 2023
Delayed puberty, gonadotropin abnormalities and subfertility in male Padi2/Padi4 double knockout mice.
Reproductive biology and endocrinology : RB&E
Sams KL,Mukai C,Marks BA,Mittal C,Demeter EA,Nelissen S,Grenier JK,Tate AE,Ahmed F,Coonrod SA
Wed Oct 12 00:00:00 EDT 2022
Conservation and divergence of myelin proteome and oligodendrocyte transcriptome profiles between humans and mice.
eLife
Gargareta VI,Reuschenbach J,Siems SB,Sun T,Piepkorn L,Mangana C,Späte E,Goebbels S,Huitinga I,Möbius W,Nave KA,Jahn O,Werner HB
Wed May 11 00:00:00 EDT 2022
Peptidylarginine deiminase enzymes and citrullinated proteins in female reproductive physiology and associated diseases†.
Biology of reproduction
Christensen AO,Li G,Young CH,Snow B,Khan SA,DeVore SB,Edwards S,Bouma GJ,Navratil AM,Cherrington BD,Rothfuss HM
Sat Dec 10 00:00:00 EST 2022
Peptidylarginine deiminase enzymes and citrullinated proteins in female reproductive physiology and associated diseases†.
Biology of reproduction
Christensen AO,Li G,Young CH,Snow B,Khan SA,DeVore SB,Edwards S,Bouma GJ,Navratil AM,Cherrington BD,Rothfuss HM
Sat Dec 10 00:00:00 EST 2022
Peptidylarginine deiminase enzymes and citrullinated proteins in female reproductive physiology and associated diseases†.
Biology of reproduction
Christensen AO,Li G,Young CH,Snow B,Khan SA,DeVore SB,Edwards S,Bouma GJ,Navratil AM,Cherrington BD,Rothfuss HM
Sat Dec 10 00:00:00 EST 2022
Peptidylarginine deiminase enzymes and citrullinated proteins in female reproductive physiology and associated diseases†.
Biology of reproduction
Christensen AO,Li G,Young CH,Snow B,Khan SA,DeVore SB,Edwards S,Bouma GJ,Navratil AM,Cherrington BD,Rothfuss HM
Sat Dec 10 00:00:00 EST 2022
Peptidylarginine deiminase enzymes and citrullinated proteins in female reproductive physiology and associated diseases†.
Biology of reproduction
Christensen AO,Li G,Young CH,Snow B,Khan SA,DeVore SB,Edwards S,Bouma GJ,Navratil AM,Cherrington BD,Rothfuss HM
Sat Dec 10 00:00:00 EST 2022
Peptidylarginine deiminase enzymes and citrullinated proteins in female reproductive physiology and associated diseases†.
Biology of reproduction
Christensen AO,Li G,Young CH,Snow B,Khan SA,DeVore SB,Edwards S,Bouma GJ,Navratil AM,Cherrington BD,Rothfuss HM
Sat Dec 10 00:00:00 EST 2022
Identification of super-enhancer-driven peptidyl arginine deiminases as potential biomarkers and therapeutic targets for osimertinib-resistant non-small cell lung cancer.
Frontiers in pharmacology
Li H,Muhetaer G,Xie Y,Yao K,Ma Q,Guan H,Xing S,Huang X,Zhou J
Sat Dec 10 00:00:00 EST 2022
IL-22 Downregulates Peptidylarginine Deiminase-1 in Human Keratinocytes: Adding Another Piece to the IL-22 Puzzle in Epidermal Barrier Formation.
The Journal of investigative dermatology
Padhi A,Srivastava A,Ramesh A,Ehrström M,Simon M,Sonkoly E,Eidsmo L,Bergman P,Lysell J
Tue Feb 01 00:00:00 EST 2022
PAD-2-mediated citrullination of nucleophosmin provides an effective target for tumor immunotherapy.
Journal for immunotherapy of cancer
Choudhury RH,Symonds P,Paston SJ,Daniels I,Cook KW,Gijon M,Metheringham RL,Brentville VA,Durrant LG
Tue Feb 01 00:00:00 EST 2022
PAD-2-mediated citrullination of nucleophosmin provides an effective target for tumor immunotherapy.
Journal for immunotherapy of cancer
Choudhury RH,Symonds P,Paston SJ,Daniels I,Cook KW,Gijon M,Metheringham RL,Brentville VA,Durrant LG
Tue Feb 01 00:00:00 EST 2022
Progesterone stimulates histone citrullination to increase IGFBP1 expression in uterine cells.
Reproduction (Cambridge, England)
Young CH,Snow B,DeVore SB,Mohandass A,Nemmara VV,Thompson PR,Thyagarajan B,Navratil AM,Cherrington BD
Thu Jul 08 00:00:00 EDT 2021
Angiogenic responses in a 3D micro-engineered environment of primary endothelial cells and pericytes.
Angiogenesis
Bai J,Khajavi M,Sui L,Fu H,Tarakkad Krishnaji S,Birsner AE,Bazinet L,Kamm RD,D'Amato RJ
Mon Feb 01 00:00:00 EST 2021
Angiogenic responses in a 3D micro-engineered environment of primary endothelial cells and pericytes.
Angiogenesis
Bai J,Khajavi M,Sui L,Fu H,Tarakkad Krishnaji S,Birsner AE,Bazinet L,Kamm RD,D'Amato RJ
Mon Feb 01 00:00:00 EST 2021
Citrullination Alters the Antiviral and Immunomodulatory Activities of the Human Cathelicidin LL-37 During Rhinovirus Infection.
Frontiers in immunology
Casanova V,Sousa FH,Shakamuri P,Svoboda P,Buch C,D'Acremont M,Christophorou MA,Pohl J,Stevens C,Barlow PG
Wed Mar 03 00:00:00 EST 2021
Citrullination Alters the Antiviral and Immunomodulatory Activities of the Human Cathelicidin LL-37 During Rhinovirus Infection.
Frontiers in immunology
Casanova V,Sousa FH,Shakamuri P,Svoboda P,Buch C,D'Acremont M,Christophorou MA,Pohl J,Stevens C,Barlow PG
Wed Mar 03 00:00:00 EST 2021
Peptidylarginine deiminase 2 has potential as both a biomarker and therapeutic target of sepsis.
JCI insight
Tian Y,Qu S,Alam HB,Williams AM,Wu Z,Deng Q,Pan B,Zhou J,Liu B,Duan X,Ma J,Mondal S,Thompson PR,Stringer KA,Standiford TJ,Li Y
Thu Oct 15 00:00:00 EDT 2020
Effect of Porphyromonas gingivalis infection on gut dysbiosis and resultant arthritis exacerbation in mouse model.
Arthritis research & therapy
Hamamoto Y,Ouhara K,Munenaga S,Shoji M,Ozawa T,Hisatsune J,Kado I,Kajiya M,Matsuda S,Kawai T,Mizuno N,Fujita T,Hirata S,Tanimoto K,Nakayama K,Kishi H,Sugiyama E,Kurihara H
Mon Oct 19 00:00:00 EDT 2020
Neutrophil extracellular traps mediate articular cartilage damage and enhance cartilage component immunogenicity in rheumatoid arthritis.
JCI insight
Carmona-Rivera C,Carlucci PM,Goel RR,James E,Brooks SR,Rims C,Hoffmann V,Fox DA,Buckner JH,Kaplan MJ
Thu Jul 09 00:00:00 EDT 2020
Peptidylarginine deiminase 4 overexpression resensitizes MCF-7/ADR breast cancer cells to adriamycin via GSK3β/p53 activation.
Cancer management and research
Zhou Q,Song C,Liu X,Qin H,Miao L,Zhang X
Thu Mar 31 00:00:00 EDT 2022
Peptidylarginine deiminase 4 overexpression resensitizes MCF-7/ADR breast cancer cells to adriamycin via GSK3β/p53 activation.
Cancer management and research
Zhou Q,Song C,Liu X,Qin H,Miao L,Zhang X
Thu Mar 31 00:00:00 EDT 2022
A Novel Citrullinated Modification of Histone 3 and Its Regulatory Mechanisms Related to IPO-38 Antibody-Labeled Protein.
Frontiers in oncology
Song S,Xiang Z,Li J,Ji J,Yan R,Zhu Z,Yu Y
Wed Sep 30 00:00:00 EDT 2020
A Novel Citrullinated Modification of Histone 3 and Its Regulatory Mechanisms Related to IPO-38 Antibody-Labeled Protein.
Frontiers in oncology
Song S,Xiang Z,Li J,Ji J,Yan R,Zhu Z,Yu Y
Wed Sep 30 00:00:00 EDT 2020
Neutrophil extracellular traps, B cells, and type I interferons contribute to immune dysregulation in hidradenitis suppurativa.
Science translational medicine
Byrd AS,Carmona-Rivera C,O'Neil LJ,Carlucci PM,Cisar C,Rosenberg AZ,Kerns ML,Caffrey JA,Milner SM,Sacks JM,Aliu O,Broderick KP,Reichner JS,Miller LS,Kang S,Robinson WH,Okoye GA,Kaplan MJ
Wed Sep 04 00:00:00 EDT 2019
Neutrophil extracellular traps, B cells, and type I interferons contribute to immune dysregulation in hidradenitis suppurativa.
Science translational medicine
Byrd AS,Carmona-Rivera C,O'Neil LJ,Carlucci PM,Cisar C,Rosenberg AZ,Kerns ML,Caffrey JA,Milner SM,Sacks JM,Aliu O,Broderick KP,Reichner JS,Miller LS,Kang S,Robinson WH,Okoye GA,Kaplan MJ
Wed Sep 04 00:00:00 EDT 2019
PAD2-Mediated Citrullination Contributes to Efficient Oligodendrocyte Differentiation and Myelination.
Cell reports
Falcão AM,Meijer M,Scaglione A,Rinwa P,Agirre E,Liang J,Larsen SC,Heskol A,Frawley R,Klingener M,Varas-Godoy M,Raposo AASF,Ernfors P,Castro DS,Nielsen ML,Casaccia P,Castelo-Branco G
Tue Apr 23 00:00:00 EDT 2019
PAD2-Mediated Citrullination Contributes to Efficient Oligodendrocyte Differentiation and Myelination.
Cell reports
Falcão AM,Meijer M,Scaglione A,Rinwa P,Agirre E,Liang J,Larsen SC,Heskol A,Frawley R,Klingener M,Varas-Godoy M,Raposo AASF,Ernfors P,Castro DS,Nielsen ML,Casaccia P,Castelo-Branco G
Tue Apr 23 00:00:00 EDT 2019
Arginine Citrullination at the C-Terminal Domain Controls RNA Polymerase II Transcription.
Molecular cell
Sharma P,Lioutas A,Fernandez-Fuentes N,Quilez J,Carbonell-Caballero J,Wright RHG,Di Vona C,Le Dily F,Schüller R,Eick D,Oliva B,Beato M
Thu Jan 03 00:00:00 EST 2019
Inhibiting PAD2 enhances the anti-tumor effect of docetaxel in tamoxifen-resistant breast cancer cells.
Journal of experimental & clinical cancer research : CR
Li F,Miao L,Xue T,Qin H,Mondal S,Thompson PR,Coonrod SA,Liu X,Zhang X
Thu Oct 10 00:00:00 EDT 2019
Modifying Enzymes Are Elicited by ER Stress, Generating Epitopes That Are Selectively Recognized by CD4+ T Cells in Patients With Type 1 Diabetes.
Diabetes
Marre ML,McGinty JW,Chow IT,DeNicola ME,Beck NW,Kent SC,Powers AC,Bottino R,Harlan DM,Greenbaum CJ,Kwok WW,Piganelli JD,James EA
Sun Jul 01 00:00:00 EDT 2018
Modifying Enzymes Are Elicited by ER Stress, Generating Epitopes That Are Selectively Recognized by CD4+ T Cells in Patients With Type 1 Diabetes.
Diabetes
Marre ML,McGinty JW,Chow IT,DeNicola ME,Beck NW,Kent SC,Powers AC,Bottino R,Harlan DM,Greenbaum CJ,Kwok WW,Piganelli JD,James EA
Sun Jul 01 00:00:00 EDT 2018
The Development of Benzimidazole-Based Clickable Probes for the Efficient Labeling of Cellular Protein Arginine Deiminases (PADs).
ACS chemical biology
Nemmara VV,Subramanian V,Muth A,Mondal S,Salinger AJ,Maurais AJ,Tilvawala R,Weerapana E,Thompson PR
Fri Mar 16 00:00:00 EDT 2018
The Development of Benzimidazole-Based Clickable Probes for the Efficient Labeling of Cellular Protein Arginine Deiminases (PADs).
ACS chemical biology
Nemmara VV,Subramanian V,Muth A,Mondal S,Salinger AJ,Maurais AJ,Tilvawala R,Weerapana E,Thompson PR
Fri Mar 16 00:00:00 EDT 2018
Activity-dependent Synaptic Recruitment of Neuroligin 1 in Spinal Dorsal Horn Contributed to Inflammatory Pain.
Neuroscience
Zhao JY,Duan XL,Yang L,Liu JP,He YT,Guo Z,Hu XD,Suo ZW
Sat Sep 15 00:00:00 EDT 2018
BB-Cl-Amidine as a novel therapeutic for canine and feline mammary cancer via activation of the endoplasmic reticulum stress pathway.
BMC cancer
Ledet MM,Anderson R,Harman R,Muth A,Thompson PR,Coonrod SA,Van de Walle GR
Thu Apr 12 00:00:00 EDT 2018
BB-Cl-Amidine as a novel therapeutic for canine and feline mammary cancer via activation of the endoplasmic reticulum stress pathway.
BMC cancer
Ledet MM,Anderson R,Harman R,Muth A,Thompson PR,Coonrod SA,Van de Walle GR
Thu Apr 12 00:00:00 EDT 2018
BB-Cl-Amidine as a novel therapeutic for canine and feline mammary cancer via activation of the endoplasmic reticulum stress pathway.
BMC cancer
Ledet MM,Anderson R,Harman R,Muth A,Thompson PR,Coonrod SA,Van de Walle GR
Thu Apr 12 00:00:00 EDT 2018
BB-Cl-Amidine as a novel therapeutic for canine and feline mammary cancer via activation of the endoplasmic reticulum stress pathway.
BMC cancer
Ledet MM,Anderson R,Harman R,Muth A,Thompson PR,Coonrod SA,Van de Walle GR
Thu Apr 12 00:00:00 EDT 2018
Histone Citrullination Represses MicroRNA Expression, Resulting in Increased Oncogene mRNAs in Somatolactotrope Cells.
Molecular and cellular biology
DeVore SB,Young CH,Li G,Sundararajan A,Ramaraj T,Mudge J,Schilkey F,Muth A,Thompson PR,Cherrington BD
Mon Oct 01 00:00:00 EDT 2018
Accumulation of citrullinated glial fibrillary acidic protein in a mouse model of bile duct ligation-induced hepatic fibrosis.
PloS one
Kim SE,Park JW,Kim MJ,Jang B,Jeon YC,Kim HJ,Ishigami A,Kim HS,Suk KT,Kim DJ,Park CK,Choi EK,Jang MK
Thu Jan 31 00:00:00 EST 2019
Accumulation of citrullinated glial fibrillary acidic protein in a mouse model of bile duct ligation-induced hepatic fibrosis.
PloS one
Kim SE,Park JW,Kim MJ,Jang B,Jeon YC,Kim HJ,Ishigami A,Kim HS,Suk KT,Kim DJ,Park CK,Choi EK,Jang MK
Thu Jan 31 00:00:00 EST 2019
Peptidylarginine deiminases 2 and 4 modulate innate and adaptive immune responses in TLR-7-dependent lupus.
JCI insight
Liu Y,Lightfoot YL,Seto N,Carmona-Rivera C,Moore E,Goel R,O'Neil L,Mistry P,Hoffmann V,Mondal S,Premnath PN,Gribbons K,Dell'Orso S,Jiang K,Thompson PR,Sun HW,Coonrod SA,Kaplan MJ
Thu Dec 06 00:00:00 EST 2018
Peptidylarginine deiminases 2 and 4 modulate innate and adaptive immune responses in TLR-7-dependent lupus.
JCI insight
Liu Y,Lightfoot YL,Seto N,Carmona-Rivera C,Moore E,Goel R,O'Neil L,Mistry P,Hoffmann V,Mondal S,Premnath PN,Gribbons K,Dell'Orso S,Jiang K,Thompson PR,Sun HW,Coonrod SA,Kaplan MJ
Thu Dec 06 00:00:00 EST 2018
Peptidylarginine deiminases 2 and 4 modulate innate and adaptive immune responses in TLR-7-dependent lupus.
JCI insight
Liu Y,Lightfoot YL,Seto N,Carmona-Rivera C,Moore E,Goel R,O'Neil L,Mistry P,Hoffmann V,Mondal S,Premnath PN,Gribbons K,Dell'Orso S,Jiang K,Thompson PR,Sun HW,Coonrod SA,Kaplan MJ
Thu Dec 06 00:00:00 EST 2018
Peptidylarginine deiminases 2 and 4 modulate innate and adaptive immune responses in TLR-7-dependent lupus.
JCI insight
Liu Y,Lightfoot YL,Seto N,Carmona-Rivera C,Moore E,Goel R,O'Neil L,Mistry P,Hoffmann V,Mondal S,Premnath PN,Gribbons K,Dell'Orso S,Jiang K,Thompson PR,Sun HW,Coonrod SA,Kaplan MJ
Thu Dec 06 00:00:00 EST 2018
PADI2-Mediated Citrullination Promotes Prostate Cancer Progression.
Cancer research
Wang L,Song G,Zhang X,Feng T,Pan J,Chen W,Yang M,Bai X,Pang Y,Yu J,Han J,Han B
Wed Nov 01 00:00:00 EDT 2017
PADI2-Mediated Citrullination Promotes Prostate Cancer Progression.
Cancer research
Wang L,Song G,Zhang X,Feng T,Pan J,Chen W,Yang M,Bai X,Pang Y,Yu J,Han J,Han B
Wed Nov 01 00:00:00 EDT 2017
PADI2-Mediated Citrullination Promotes Prostate Cancer Progression.
Cancer research
Wang L,Song G,Zhang X,Feng T,Pan J,Chen W,Yang M,Bai X,Pang Y,Yu J,Han J,Han B
Wed Nov 01 00:00:00 EDT 2017
PADI2-Mediated Citrullination Promotes Prostate Cancer Progression.
Cancer research
Wang L,Song G,Zhang X,Feng T,Pan J,Chen W,Yang M,Bai X,Pang Y,Yu J,Han J,Han B
Wed Nov 01 00:00:00 EDT 2017
PADI2-Mediated Citrullination Promotes Prostate Cancer Progression.
Cancer research
Wang L,Song G,Zhang X,Feng T,Pan J,Chen W,Yang M,Bai X,Pang Y,Yu J,Han J,Han B
Wed Nov 01 00:00:00 EDT 2017
PADI2-Mediated Citrullination Promotes Prostate Cancer Progression.
Cancer research
Wang L,Song G,Zhang X,Feng T,Pan J,Chen W,Yang M,Bai X,Pang Y,Yu J,Han J,Han B
Wed Nov 01 00:00:00 EDT 2017
PADI2-Mediated Citrullination Promotes Prostate Cancer Progression.
Cancer research
Wang L,Song G,Zhang X,Feng T,Pan J,Chen W,Yang M,Bai X,Pang Y,Yu J,Han J,Han B
Wed Nov 01 00:00:00 EDT 2017
Protein-arginine deiminase 2 suppresses proliferation of colon cancer cells through protein citrullination.
Cancer science
Funayama R,Taniguchi H,Mizuma M,Fujishima F,Kobayashi M,Ohnuma S,Unno M,Nakayama K
Sat Apr 01 00:00:00 EDT 2017
Protein-arginine deiminase 2 suppresses proliferation of colon cancer cells through protein citrullination.
Cancer science
Funayama R,Taniguchi H,Mizuma M,Fujishima F,Kobayashi M,Ohnuma S,Unno M,Nakayama K
Sat Apr 01 00:00:00 EDT 2017
Protein-arginine deiminase 2 suppresses proliferation of colon cancer cells through protein citrullination.
Cancer science
Funayama R,Taniguchi H,Mizuma M,Fujishima F,Kobayashi M,Ohnuma S,Unno M,Nakayama K
Sat Apr 01 00:00:00 EDT 2017
Protein-arginine deiminase 2 suppresses proliferation of colon cancer cells through protein citrullination.
Cancer science
Funayama R,Taniguchi H,Mizuma M,Fujishima F,Kobayashi M,Ohnuma S,Unno M,Nakayama K
Sat Apr 01 00:00:00 EDT 2017
Citrullination regulates the expression of insulin-like growth factor-binding protein 1 (IGFBP1) in ovine uterine luminal epithelial cells.
Reproduction (Cambridge, England)
Young CH,Rothfuss HM,Gard PF,Muth A,Thompson PR,Ashley RL,Cherrington BD
Sun Jan 01 00:00:00 EST 2017
The lncRNA HOXA11-AS functions as a competing endogenous RNA to regulate PADI2 expression by sponging miR-125a-5p in liver metastasis of colorectal cancer.
Oncotarget
Chen D,Sun Q,Zhang L,Zhou X,Cheng X,Zhou D,Ye F,Lin J,Wang W
Tue Sep 19 00:00:00 EDT 2017
Role of peptidylarginine deiminase 2 (PAD2) in mammary carcinoma cell migration.
BMC cancer
Horibata S,Rogers KE,Sadegh D,Anguish LJ,McElwee JL,Shah P,Thompson PR,Coonrod SA
Fri May 26 00:00:00 EDT 2017
Role of peptidylarginine deiminase 2 (PAD2) in mammary carcinoma cell migration.
BMC cancer
Horibata S,Rogers KE,Sadegh D,Anguish LJ,McElwee JL,Shah P,Thompson PR,Coonrod SA
Fri May 26 00:00:00 EDT 2017
Role of peptidylarginine deiminase 2 (PAD2) in mammary carcinoma cell migration.
BMC cancer
Horibata S,Rogers KE,Sadegh D,Anguish LJ,McElwee JL,Shah P,Thompson PR,Coonrod SA
Fri May 26 00:00:00 EDT 2017
Role of peptidylarginine deiminase 2 (PAD2) in mammary carcinoma cell migration.
BMC cancer
Horibata S,Rogers KE,Sadegh D,Anguish LJ,McElwee JL,Shah P,Thompson PR,Coonrod SA
Fri May 26 00:00:00 EDT 2017
A cellular model reflecting the phenotypic heterogeneity of mutant HRAS driven squamous cell carcinoma.
International journal of cancer
Cantariño N,Fernández-Figueras MT,Valero V,Musulén E,Malinverni R,Granada I,Goldie SJ,Martín-Caballero J,Douet J,Forcales SV,Buschbeck M
Thu Sep 01 00:00:00 EDT 2016
A cellular model reflecting the phenotypic heterogeneity of mutant HRAS driven squamous cell carcinoma.
International journal of cancer
Cantariño N,Fernández-Figueras MT,Valero V,Musulén E,Malinverni R,Granada I,Goldie SJ,Martín-Caballero J,Douet J,Forcales SV,Buschbeck M
Thu Sep 01 00:00:00 EDT 2016
Downregulation of the Deiminase PADI2 Is an Early Event in Colorectal Carcinogenesis and Indicates Poor Prognosis.
Molecular cancer research : MCR
Cantariño N,Musulén E,Valero V,Peinado MA,Perucho M,Moreno V,Forcales SV,Douet J,Buschbeck M
Thu Sep 01 00:00:00 EDT 2016
Downregulation of the Deiminase PADI2 Is an Early Event in Colorectal Carcinogenesis and Indicates Poor Prognosis.
Molecular cancer research : MCR
Cantariño N,Musulén E,Valero V,Peinado MA,Perucho M,Moreno V,Forcales SV,Douet J,Buschbeck M
Thu Sep 01 00:00:00 EDT 2016
Significant impact of miRNA-target gene networks on genetics of human complex traits.
Scientific reports
Okada Y,Muramatsu T,Suita N,Kanai M,Kawakami E,Iotchkova V,Soranzo N,Inazawa J,Tanaka T
Tue Mar 01 00:00:00 EST 2016
Significant impact of miRNA-target gene networks on genetics of human complex traits.
Scientific reports
Okada Y,Muramatsu T,Suita N,Kanai M,Kawakami E,Iotchkova V,Soranzo N,Inazawa J,Tanaka T
Tue Mar 01 00:00:00 EST 2016
Peptidylarginine deiminase 1-catalyzed histone citrullination is essential for early embryo development.
Scientific reports
Zhang X,Liu X,Zhang M,Li T,Muth A,Thompson PR,Coonrod SA,Zhang X
Thu Dec 08 00:00:00 EST 2016
Peptidylarginine deiminase 1-catalyzed histone citrullination is essential for early embryo development.
Scientific reports
Zhang X,Liu X,Zhang M,Li T,Muth A,Thompson PR,Coonrod SA,Zhang X
Thu Dec 08 00:00:00 EST 2016
GnRH Stimulates Peptidylarginine Deiminase Catalyzed Histone Citrullination in Gonadotrope Cells.
Molecular endocrinology (Baltimore, Md.)
Khan SA,Edwards BS,Muth A,Thompson PR,Cherrington BD,Navratil AM
Sat Oct 01 00:00:00 EDT 2016
GnRH Stimulates Peptidylarginine Deiminase Catalyzed Histone Citrullination in Gonadotrope Cells.
Molecular endocrinology (Baltimore, Md.)
Khan SA,Edwards BS,Muth A,Thompson PR,Cherrington BD,Navratil AM
Sat Oct 01 00:00:00 EDT 2016
GnRH Stimulates Peptidylarginine Deiminase Catalyzed Histone Citrullination in Gonadotrope Cells.
Molecular endocrinology (Baltimore, Md.)
Khan SA,Edwards BS,Muth A,Thompson PR,Cherrington BD,Navratil AM
Sat Oct 01 00:00:00 EDT 2016
GnRH Stimulates Peptidylarginine Deiminase Catalyzed Histone Citrullination in Gonadotrope Cells.
Molecular endocrinology (Baltimore, Md.)
Khan SA,Edwards BS,Muth A,Thompson PR,Cherrington BD,Navratil AM
Sat Oct 01 00:00:00 EDT 2016
Decreased severity of experimental autoimmune arthritis in peptidylarginine deiminase type 4 knockout mice.
BMC musculoskeletal disorders
Suzuki A,Kochi Y,Shoda H,Seri Y,Fujio K,Sawada T,Yamada R,Yamamoto K
Thu May 05 00:00:00 EDT 2016
Decreased severity of experimental autoimmune arthritis in peptidylarginine deiminase type 4 knockout mice.
BMC musculoskeletal disorders
Suzuki A,Kochi Y,Shoda H,Seri Y,Fujio K,Sawada T,Yamada R,Yamamoto K
Thu May 05 00:00:00 EDT 2016
Chemical Proteomic Platform To Identify Citrullinated Proteins.
ACS chemical biology
Lewallen DM,Bicker KL,Subramanian V,Clancy KW,Slade DJ,Martell J,Dreyton CJ,Sokolove J,Weerapana E,Thompson PR
Fri Nov 20 00:00:00 EST 2015
Chemical Proteomic Platform To Identify Citrullinated Proteins.
ACS chemical biology
Lewallen DM,Bicker KL,Subramanian V,Clancy KW,Slade DJ,Martell J,Dreyton CJ,Sokolove J,Weerapana E,Thompson PR
Fri Nov 20 00:00:00 EST 2015
PAD2 overexpression in transgenic mice promotes spontaneous skin neoplasia.
Cancer research
McElwee JL,Mohanan S,Horibata S,Sams KL,Anguish LJ,McLean D,Cvitaš I,Wakshlag JJ,Coonrod SA
Sat Nov 01 00:00:00 EDT 2014
PAD2 overexpression in transgenic mice promotes spontaneous skin neoplasia.
Cancer research
McElwee JL,Mohanan S,Horibata S,Sams KL,Anguish LJ,McLean D,Cvitaš I,Wakshlag JJ,Coonrod SA
Sat Nov 01 00:00:00 EDT 2014
PAD2 overexpression in transgenic mice promotes spontaneous skin neoplasia.
Cancer research
McElwee JL,Mohanan S,Horibata S,Sams KL,Anguish LJ,McLean D,Cvitaš I,Wakshlag JJ,Coonrod SA
Sat Nov 01 00:00:00 EDT 2014
PAD2 overexpression in transgenic mice promotes spontaneous skin neoplasia.
Cancer research
McElwee JL,Mohanan S,Horibata S,Sams KL,Anguish LJ,McLean D,Cvitaš I,Wakshlag JJ,Coonrod SA
Sat Nov 01 00:00:00 EDT 2014
PAD2 overexpression in transgenic mice promotes spontaneous skin neoplasia.
Cancer research
McElwee JL,Mohanan S,Horibata S,Sams KL,Anguish LJ,McLean D,Cvitaš I,Wakshlag JJ,Coonrod SA
Sat Nov 01 00:00:00 EDT 2014
Targeted H3R26 deimination specifically facilitates estrogen receptor binding by modifying nucleosome structure.
PLoS genetics
Guertin MJ,Zhang X,Anguish L,Kim S,Varticovski L,Lis JT,Hager GL,Coonrod SA
Mon Sep 01 00:00:00 EDT 2014
Targeted H3R26 deimination specifically facilitates estrogen receptor binding by modifying nucleosome structure.
PLoS genetics
Guertin MJ,Zhang X,Anguish L,Kim S,Varticovski L,Lis JT,Hager GL,Coonrod SA
Mon Sep 01 00:00:00 EDT 2014
Identification of macrophage extracellular trap-like structures in mammary gland adipose tissue: a preliminary study.
Frontiers in immunology
Mohanan S,Horibata S,McElwee JL,Dannenberg AJ,Coonrod SA
Wed Mar 20 00:00:00 EDT 2013
Identification of macrophage extracellular trap-like structures in mammary gland adipose tissue: a preliminary study.
Frontiers in immunology
Mohanan S,Horibata S,McElwee JL,Dannenberg AJ,Coonrod SA
Wed Mar 20 00:00:00 EDT 2013
Peptidylarginine deiminase 2-catalyzed histone H3 arginine 26 citrullination facilitates estrogen receptor α target gene activation.
Proceedings of the National Academy of Sciences of the United States of America
Zhang X,Bolt M,Guertin MJ,Chen W,Zhang S,Cherrington BD,Slade DJ,Dreyton CJ,Subramanian V,Bicker KL,Thompson PR,Mancini MA,Lis JT,Coonrod SA
Tue Aug 14 00:00:00 EDT 2012
Peptidylarginine deiminase 2-catalyzed histone H3 arginine 26 citrullination facilitates estrogen receptor α target gene activation.
Proceedings of the National Academy of Sciences of the United States of America
Zhang X,Bolt M,Guertin MJ,Chen W,Zhang S,Cherrington BD,Slade DJ,Dreyton CJ,Subramanian V,Bicker KL,Thompson PR,Mancini MA,Lis JT,Coonrod SA
Tue Aug 14 00:00:00 EDT 2012
ATP induces protein arginine deiminase 2-dependent citrullination in mast cells through the P2X7 purinergic receptor.
Journal of immunology (Baltimore, Md. : 1950)
Arandjelovic S,McKenney KR,Leming SS,Mowen KA
Mon Oct 15 00:00:00 EDT 2012
ATP induces protein arginine deiminase 2-dependent citrullination in mast cells through the P2X7 purinergic receptor.
Journal of immunology (Baltimore, Md. : 1950)
Arandjelovic S,McKenney KR,Leming SS,Mowen KA
Mon Oct 15 00:00:00 EDT 2012
Comparative analysis of peptidylarginine deiminase-2 expression in canine, feline and human mammary tumours.
Journal of comparative pathology
Cherrington BD,Mohanan S,Diep AN,Fleiss R,Sudilovsky D,Anguish LJ,Coonrod SA,Wakshlag JJ
Tue Jan 22 00:00:00 EST 2013
Comparative analysis of peptidylarginine deiminase-2 expression in canine, feline and human mammary tumours.
Journal of comparative pathology
Cherrington BD,Mohanan S,Diep AN,Fleiss R,Sudilovsky D,Anguish LJ,Coonrod SA,Wakshlag JJ
Tue Jan 22 00:00:00 EST 2013
Potential role for PAD2 in gene regulation in breast cancer cells.
PloS one
Cherrington BD,Zhang X,McElwee JL,Morency E,Anguish LJ,Coonrod SA
Fri Apr 19 00:00:00 EDT 2013
Potential role for PAD2 in gene regulation in breast cancer cells.
PloS one
Cherrington BD,Zhang X,McElwee JL,Morency E,Anguish LJ,Coonrod SA
Fri Apr 19 00:00:00 EDT 2013
Potential role for PAD2 in gene regulation in breast cancer cells.
PloS one
Cherrington BD,Zhang X,McElwee JL,Morency E,Anguish LJ,Coonrod SA
Fri Apr 19 00:00:00 EDT 2013
Potential role for PAD2 in gene regulation in breast cancer cells.
PloS one
Cherrington BD,Zhang X,McElwee JL,Morency E,Anguish LJ,Coonrod SA
Fri Apr 19 00:00:00 EDT 2013
Potential role for PAD2 in gene regulation in breast cancer cells.
PloS one
Cherrington BD,Zhang X,McElwee JL,Morency E,Anguish LJ,Coonrod SA
Fri Apr 19 00:00:00 EDT 2013
Identification of PADI2 as a potential breast cancer biomarker and therapeutic target.
BMC cancer
McElwee JL,Mohanan S,Griffith OL,Breuer HC,Anguish LJ,Cherrington BD,Palmer AM,Howe LR,Subramanian V,Causey CP,Thompson PR,Gray JW,Coonrod SA
Tue Oct 30 00:00:00 EDT 2012
Identification of PADI2 as a potential breast cancer biomarker and therapeutic target.
BMC cancer
McElwee JL,Mohanan S,Griffith OL,Breuer HC,Anguish LJ,Cherrington BD,Palmer AM,Howe LR,Subramanian V,Causey CP,Thompson PR,Gray JW,Coonrod SA
Tue Oct 30 00:00:00 EDT 2012
Identification of PADI2 as a potential breast cancer biomarker and therapeutic target.
BMC cancer
McElwee JL,Mohanan S,Griffith OL,Breuer HC,Anguish LJ,Cherrington BD,Palmer AM,Howe LR,Subramanian V,Causey CP,Thompson PR,Gray JW,Coonrod SA
Tue Oct 30 00:00:00 EDT 2012
Identification of PADI2 as a potential breast cancer biomarker and therapeutic target.
BMC cancer
McElwee JL,Mohanan S,Griffith OL,Breuer HC,Anguish LJ,Cherrington BD,Palmer AM,Howe LR,Subramanian V,Causey CP,Thompson PR,Gray JW,Coonrod SA
Tue Oct 30 00:00:00 EDT 2012
Genome-wide analysis reveals PADI4 cooperates with Elk-1 to activate c-Fos expression in breast cancer cells.
PLoS genetics
Zhang X,Gamble MJ,Stadler S,Cherrington BD,Causey CP,Thompson PR,Roberson MS,Kraus WL,Coonrod SA
Wed Jun 01 00:00:00 EDT 2011
Genome-wide analysis reveals PADI4 cooperates with Elk-1 to activate c-Fos expression in breast cancer cells.
PLoS genetics
Zhang X,Gamble MJ,Stadler S,Cherrington BD,Causey CP,Thompson PR,Roberson MS,Kraus WL,Coonrod SA
Wed Jun 01 00:00:00 EDT 2011
Potential role for peptidylarginine deiminase 2 (PAD2) in citrullination of canine mammary epithelial cell histones.
PloS one
Cherrington BD,Morency E,Struble AM,Coonrod SA,Wakshlag JJ
Mon Jul 26 00:00:00 EDT 2010
Potential role for peptidylarginine deiminase 2 (PAD2) in citrullination of canine mammary epithelial cell histones.
PloS one
Cherrington BD,Morency E,Struble AM,Coonrod SA,Wakshlag JJ
Mon Jul 26 00:00:00 EDT 2010
Potential role for peptidylarginine deiminase 2 (PAD2) in citrullination of canine mammary epithelial cell histones.
PloS one
Cherrington BD,Morency E,Struble AM,Coonrod SA,Wakshlag JJ
Mon Jul 26 00:00:00 EDT 2010