Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSLQRIVRVS LEHPTSAVCV AGVETLVDIY GSVPEGTEMF EVYGTPGVDI YISPNMERGR ERADTRRWRF DATLEIIVVM NSPSNDLNDS HVQISYHSSH EPLPLAYAVL YLTCVDISLD CDLNCEGRQD RNFVDKRQWV WGPSGYGGIL LVNCDRDDPS CDVQDNCDQH VHCLQDLEDM SVMVLRTQGP AALFDDHKLV LHTSSYDAKR AQVFHICGPE DVCEAYRHVL GQDKVSYEVP RLHGDEERFF VEGLSFPDAG FTGLISFHVT LLDDSNEDFS ASPIFTDTVV FRVAPWIMTP STLPPLEVYV CRVRNNTCFV DAVAELARKA GCKLTICPQA ENRNDRWIQD EMELGYVQAP HKTLPVVFDS PRNGELQDFP YKRILGPDFG YVTREPRDRS VSGLDSFGNL EVSPPVVANG KEYPLGRILI GGNLPGSSGR RVTQVVRDFL HAQKVQPPVE LFVDWLAVGH VDEFLSFVPA PDGKGFRMLL ASPGACFKLF QEKQKCGHGR ALLFQGVVDD EQVKTISINQ VLSNKDLINY NKFVQSCIDW NREVLKRELG LAECDIIDIP QLFKTERKKA TAFFPDLVNM LVLGKHLGIP KPFGPIINGC CCLEEKVRSL LEPLGLHCTF IDDFTPYHML HGEVHCGTNV CRKPFSFKWW NMVP
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: peptidyl arginine deiminase, type III; Peptidylarginine deiminase III; Protein-arginine deiminase type III; Protein-arginine deiminase type-3
Gene Aliases: PAD3; PADI3; PDI3
UniProt ID: (Human) Q9ULW8
Entrez Gene ID: (Human) 51702
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support