Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For ICC, formaldehyde fixation is recommended. Permeabilization with Triton-X 100 is recommended for formaldehyde-fixed cells. For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections.
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase I which synthesizes ribosomal RNA precursors. Isoform 1 is involved in UBTF-activated transcription, presumably at a step following PIC formation.; Isoform 2 has been described as a component of preformed T-cell receptor (TCR) complex. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A34.5; antis; Antisense to ERCC-1 protein; ASE-1; CD3-epsilon-associated protein; CD3e antigen, epsilon polypeptide associated protein; CD3e molecule, epsilon associated protein; CD3E-associated protein; DNA-directed RNA polymerase I subunit G; DNA-directed RNA polymerase I subunit RPA34; RNA polymerase I-associated factor PAF49
Gene Aliases: ASE-1; ASE1; CAST; CD3EAP; PAF49; POLR1G
UniProt ID: (Human) O15446
Entrez Gene ID: (Human) 10849
Molecular Function: DNA-directed RNA polymerase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support