Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This gene encodes an acetylhydrolase that catalyzes the removal of an acetyl group from the glycerol backbone of platelet-activating factor. The encoded enzyme is a subunit of the platelet-activating factor acetylhydrolase isoform 1B complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PAF acetylhydrolase 29 kDa subunit; PAF-AH 29 kDa subunit; PAF-AH subunit gamma; PAF-AH1b alpha 1 subunit; PAFAH subunit gamma; platelet-activating factor acetylhydrolase 1b, catalytic subunit 3 (29kDa); Platelet-activating factor acetylhydrolase IB subunit alpha1; platelet-activating factor acetylhydrolase, isoform Ib, subunit 3 (29kDa)
Gene Aliases: PAFAH1B3; PAFAHG
UniProt ID: (Human) Q15102
Entrez Gene ID: (Human) 5050
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support