Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGVNQSVGFP PVTGPHLVGC GDVMEGQNLQ GSFFRLFYPC QKAEETMEQP LWIPRYEYCT GLAEYLQFNK RCGGLLFNLA VGSCRLPVSW NGPFKTKDSG YPLIIFSHGL GAFRTLYSAF CMELASRGFV VAVPEHRDRS AATTYFCKQA PEENQPTNES LQEEWIPFRR VEEGEKEFHV RNPQVHQRVS ECLRVLKILQ EVTAGQTVFN ILPGGLDLMT LKGNIDMSRV AVMGHSFGGA TAILALAKET QFRCAVALDA WMFPLERDFY PKARGPVFFI NTEKFQTMES VNLMKKICAQ HEQSRIITVL GSVHRSQTDF AFVTGNLIGK FFSTETRGSL DPYEGQEVMV RAMLAFLQKH LDLKEDYNQW NNLIEGIGPS LTPGAPHHLS SL
Has a marked selectivity for phospholipids with short acyl chains at the sn-2 position. May share a common physiologic function with the plasma-type enzyme.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PAF:lysophospholipid transacetylase; PAF:sphingosine transacetylase; platelet-activating factor acetylhydrolase 2, 40kDa; Platelet-activating factor acetylhydrolase 2, cytoplasmic; Platelet-activating factor acetyltransferase PAFAH2; SD-PLA2; Serine-dependent phospholipase A2
Gene Aliases: HSD-PLA2; PAFAH2
UniProt ID: (Human) Q99487
Entrez Gene ID: (Human) 5051
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support