Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody has been tested in direct-ELISA
The target protein PAG1B appears to be proteolytically inactive. It is expressed in trophoblast and placental tissue. It is produced specifically in the invasive binucleate cells of the placenta. It becomes detectable in maternal serum soon after implantation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PAG 1; Pregnancy-associated glycoprotein 1; Pregnancy-specific protein B; PSP-B
Gene Aliases: PAG1B
UniProt ID: (Bovine) Q29432
Entrez Gene ID: (Bovine) 281964
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support