Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a protein containing three zinc finger domains and a nuclear localization signal. The mRNA and the protein of this gene are upregulated by wildtype p53 and overexpression of this gene inhibits tumor cell growth, suggesting that this gene may have a role in the p53-dependent growth regulatory pathway. Alternative splicing of this gene results in two transcript variants encoding two isoforms differing in only one amino acid.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ12296; matrin type 3; MGC10613; p53 target zinc finger protein; p53-activated gene 608 protein; WIG-1/PAG608 protein; wild-type p53-induced gene 1; Zinc finger matrin-type protein 3; Zinc finger protein WIG-1; zinc finger protein WIG1; zmat3 {ECO:0000250|UniProtKB:Q9HA38}
Gene Aliases: PAG608; WIG-1; WIG1; ZMAT3
UniProt ID: (Human) Q9HA38, (Rat) O08781
Entrez Gene ID: (Human) 64393, (Rat) 64394
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support