Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: MATAEVLNIG KKLYEGKTKE VYELLDSPGK VLLQSKDQIT AGNAARKNHL EGKAAISNKI TSCIFQLLQE AGIKTAFTRK CGETAFIAPQ CEMIPIEWVC RRIATGSFLK RNPGVKEGYK FYPPKVELFF KDDANNDPQW SEEQLIAAKF CFAGLLIGQT EVDIMSHATQ AIFEILEKSW LPQNCTLVDM KIEFGVDVTT KEIVLADVID NDSWRLWPSG DRSQQKDKQS YRDLKEVTPE GLQMVKKNFE WVAERVELLL KSESQCRVVV LMGSTSDLGH CEKIKKACGN FGIPCELRVT SAHKGPDETL RIKAEYEGDG IPTVFVAVAG RSNGLGPVMS GNTAYPVISC PPLTPDWGVQ DVWSSLRLPS GLGCSTVLSP EGSAQFAAQI FGLSNHLVWS KLRASILNTW ISLKQADKKI RECNL
This gene encodes a bifunctional enzyme containing phosphoribosylaminoimidazole carboxylase activity in its N-terminal region and phosphoribosylaminoimidazole succinocarboxamide synthetase in its C-terminal region. It catalyzes steps 6 and 7 of purine biosynthesis. The gene is closely linked and divergently transcribed with a locus that encodes an enzyme in the same pathway, and transcription of the two genes is coordinately regulated. The human genome contains several pseudogenes of this gene. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AIR carboxylase; Bifunctional phosphoribosylaminoimidazole carboxylase/phosphoribosylaminoimidazole succinocarboxamide synthetase; multifunctional protein ADE2H1; PAICS; phosphoribosylaminoimidazole carboxylase, phosphoribosylaminoimidazole succinocarboxamide synthetase; SAICAR synthetase
Gene Aliases: ADE2; ADE2H1; AIRC; PAICS; PAIS
UniProt ID: (Human) P22234
Entrez Gene ID: (Human) 10606
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support