Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
PAK1 is a serine/threonine kinase that is a target for restenosis, cancer, and immunosuppression treatment. These proteins serve as targets for the small GTP binding proteins Cdc42 and Rac and have been implicated in a wide range of biological activities. PAK1 regulates cell motility and morphology.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADRB2; alpha pak; Alpha-PAK; EC 2.7.11.1; kinase PAK1; MGC130000; MGC130001; p21 (CDKN1A)-activated kinase 1; p21-activated kinase 1; p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related); p68-PAK; PAK 1; PAK-1; PAK1B; Protein kinase MUK2; Serine/threonine-protein kinase PAK 1
Gene Aliases: Pak1
UniProt ID: (Rat) P35465
Entrez Gene ID: (Rat) 29431
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support