Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VEKDKVTGET RVLSSTTLLP RQPLPLGIKV YEDETKVVHA VDGTAENGIH PLSSSEVDEL IHKADEVTLS EAGSTAGAAE TRGAVEGAAR TTPSRREITG VQAQPGEAT
This gene encodes a member of the paralemmin protein family. The product of this gene is a prenylated and palmitoylated phosphoprotein that associates with the cytoplasmic face of plasma membranes and is implicated in plasma membrane dynamics in neurons and other cell types. Several alternatively spliced transcript variants have been identified, but the full-length nature of only two transcript variants has been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Paralemmin; Paralemmin-1
Gene Aliases: KIAA0270; PALM
UniProt ID: (Human) O75781
Entrez Gene ID: (Human) 5064
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support