Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Predicted reactivity by homology: Mouse.
Regulates ATP-dependent protein translocation into the mitochondrial matrix. Inhibits DNAJC19 stimulation of HSPA9/Mortalin ATPase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: magmas-like protein; mitochondria associated protein involved in granulocyte macrophage colony stimulating factor signal transduction; Mitochondria-associated granulocyte macrophage CSF-signaling molecule; Mitochondrial import inner membrane translocase subunit TIM16; Presequence translocated-associated motor subunit PAM16
Gene Aliases: CGI-136; MAGMAS; PAM16; SMDMDM; TIM16; TIMM16
UniProt ID: (Human) Q9Y3D7
Entrez Gene ID: (Human) 51025
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support