Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ATVVHILDS RTGGSLVAHI KVGETYHQRK EGVTHQQWYL FNDFLIEPID KHEAVQFDMN WKVPAILYYV KRNLNSRYNL NIKNPIEASV LLAEASLARK QRKTHTTFIP LMLNEMPQIG DLVGLDAEFV TLNEEEAELR SDGTKSTIKP SQMSVARITC VRGQGPNEGI PFIDDYISTQ EQVVDYLTQY SGIKPGDLDA KISSKHLTTL KSTYLKLRFL IDIGVKFVGH GLQKDFRVIN LMVPKDQVLD TVYLFHMPRK RMISLRFLAW YFLDLKIQGE THDSIEDART ALQLYRKYLE LSKNGTEPES FHKVLKGLYE KGRKMDWKVP EPEGQTSPKN AAVFSSVLAL (850-1198 aa encoded by BC024043)
This gene encodes a coregulator for the alpha and beta estrogen receptors and the orphan nuclear receptor NR1D2. The protein localizes to the nucleus, and is thought to have both coactivator and corepressor functions. Its interaction with nuclear receptors is independent of the AF2 domain on the receptors, which is known to regulate interaction with other coreceptors. Two alternatively spliced transcript variants for this gene have been described. However, the full length nature of one of the variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AN2 polyA specific ribonuclease subunit homolog; Inactive ubiquitin carboxyl-terminal hydrolase 52; inactive ubiquitin carboxyl-terminal hydrolase 52 {ECO:0000255|HAMAP-Rule:MF_03182}; PAB-dependent poly(A)-specific ribonuclease subunit 2; PAB-dependent poly(A)-specific ribonuclease subunit PAN2; PAB-dependent poly(A)-specific ribonuclease subunit PAN2 {ECO:0000255|HAMAP-Rule:MF_03182}; PAB1P-dependent poly(A)-nuclease; PAB1P-dependent poly(A)-nuclease {ECO:0000255|HAMAP-Rule:MF_03182}; PAB1P-dependent poly(A)-specific ribonuclease; PABP-dependent poly(A) nuclease 2; PAN deadenylation complex catalytic subunit 2; PAN deadenylation complex catalytic subunit 2 {ECO:0000255|HAMAP-Rule:MF_03182}; PAN deadenylation complex subunit 2; PAN2 poly(A) specific ribonuclease subunit homolog; PAN2 polyA specific ribonuclease subunit; PAN2 polyA specific ribonuclease subunit homolog; PAN2-PAN3 deadenylation complex catalytic subunit PAN2; Poly(A)-nuclease deadenylation complex subunit 2; ubiquitin specific peptidase 52; ubiquitin specific protease 52
Gene Aliases: 1200014O24Rik; AI047843; AW742773; KIAA0710; mKIAA0710; PAN2; USP52
UniProt ID: (Human) Q504Q3, (Mouse) Q3TZK5
Entrez Gene ID: (Human) 9924, (Rat) 408200, (Mouse) 103135
Molecular Function:
exoribonuclease
RNA metabolism protein
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support