Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MKIKDAKKPS FPWFGMDIGG TLVKLSYFEP IDITAEEEQE EVESLKSIRK YLTSNVAYGS TGIRDVHLEL KDLTLFGRRG NLHFIRFPTQ DLPTFIQMGR DKNFSTLQTV LCATGGGAYK FEKDFRTIGN LHLHKLDELD CLVKGLLYID SVSFNGQAEC YYFANASEPE RCQKMPFNLD DPYPLLVVNI GSGVSILAVH SKDNYKRVTG TSLGGGTFLG LCSLLTGCES FEEALEMASK GDSTQADKLV RDIYGGDYER FGLPGWAVAS SFGNMIYKEK RESVSKEDLA RATLVTITNN IGSVARMCAV NEKINRVVFV GNFLRVNTLS MKLLAYALDY WSKGQLKALF LEHEGYFGAV GALLGLPNFS
This gene encodes a protein belonging to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. This family member is expressed most abundantly in the liver.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPanK3; Pantothenate kinase 3; Pantothenic acid kinase 3
Gene Aliases: PANK3
UniProt ID: (Human) Q9H999
Entrez Gene ID: (Human) 79646
Molecular Function: kinase transferase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support