Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MESTGSVGEA PGGPRVLVVG GGIAGLGAAQ RLCGHSAFPH LRVLEATARA GGRIRSERCF GGVVEVGAHW IHGPSRGNPV FQLAAEYGLL GEKELSQENQ LVETGGHVGL PSVSYASSGA SVSLQLVAEM ATLFYGLIDQ TREFLHAAET PVPSVGEYLK KEIGQHVAGW TEDEETRKLK LAVLNSFFNL ECCVSGTHSM DLVALAPFGE YTVLPGLDCT FSKGYQGLTN CMMAALPEDT VVFEKPVKTI HWNGSFQEAA FPGETFPVSV ECEDGDRFPA HHVIVTVPLG FLREHLDTFF DPPLPAEKAE AIRKIGFGTN NKIFLEFEEP FWEPDCQLIQ LVWEDTSPLE DAAPELQDAW FRKLIGFVVL PAFASVHVLC GFIAGLESEF METLSDEEVL LCLTQVLRRV TGNPRLPAPK SVLRSRWHSA PYTRGSYSYV AVGSTGGDLD LLAQPLPADG AGAQLQILFA GEATHRTFYS TTHGALLSGW READRLLSLW APQVQQPRPR L
Flavoenzyme which catalyzes the oxidation of N(1)-acetylspermine to spermidine and is thus involved in the polyamine back-conversion (PubMed:12477380). Can also oxidize N(1)-acetylspermidine to putrescine. Substrate specificity: N(1)-acetylspermine = N(1)-acetylspermidine > N(1),N(12)-diacylspermine >> spermine. Does not oxidize spermidine. Plays an important role in the regulation of polyamine intracellular concentration and has the potential to act as a determinant of cellular sensitivity to the antitumor polyamine analogs (PubMed:12477380). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Peroxisomal N(1)-acetyl-spermine/spermidine oxidase; peroxisomal N1-acetyl-spermine/spermidine oxidase; Polyamine oxidase
Gene Aliases: PAO; PAOX; UNQ1923/PRO4398
UniProt ID: (Human) Q6QHF9
Entrez Gene ID: (Human) 196743
Molecular Function:
oxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support