Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RERDYAEIQD FHRTFGCDDE LMYGGVSSYE GSMALNARPQ SPREGHMMDA LYAQVKKPRN SKPSPVDSNR STPSNHDRIQ RLRQEFQQAK QDEDVEDRRR TYSFEQPWPN ARPATQSGRH SVSVEVQMQR QRQEERESSQ QAQRQYSSLP RQSRKNASSV SQDSWEQNYS PGEGFQSAKE NPRYSSYQGS RNGYLGGHGF NARVMLETQE LLRQEQRRKE QQMKKQPPSE GPSNYDSYKK VQDPSYAPPK GPFRQDVPPS PSQVARLNRL QTPEKGRPFY S
This gene encodes a member of the kinesin 4 or subfamily of kinesin related proteins. This subfamily is also referred to as the chromokinesins. The encoded protein is an ATP dependent microtubule-based motor protein that is involved in the intracellular transport of membranous organelles. This protein also associates with condensed chromosome arms and may be involved maintaining chromosome integrity during mitosis. This protein may also be involved in the organization of the central spindle prior to cytokinesis. A pseudogene of this gene is found on chromosome X.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ASIP; atypical PKC isotype-specific interacting protein; Atypical PKC isotype-specific-interacting protein; bazooka; CTCL tumor antigen se2-5; FLJ21015; PAR-3; par-3 family cell polarity regulator alpha; par-3 partitioning defective 3 homolog; PAR3-alpha; Partitioning defective 3 homolog; protein phosphatase 1, regulatory subunit 118; RP11-406D17.1
Gene Aliases: ASIP; Baz; PAR3; PAR3A; PAR3alpha; PARD-3; PARD3; PARD3A; PPP1R118; SE2-5L16; SE2-5LT1; SE2-5T2
UniProt ID: (Human) Q9NWL4
Entrez Gene ID: (Human) 56288
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support