Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MFHKAEELF SKTTNNEVDD MDTSDTQWGW FYLAECGKWH MFQPDTNSQC SVSSEDIEKS FKTNPCGSIS FTTSKFSYKI DFAEMKQMNL TTGKQRLIKR APFSISAFSY ICENEAIPMP PHWENVNTQV PYQLIPLHNQ THEYNEVANL FGKTMDRNRI KRIQRIQNLD LWEFFCRKKA QLKKKRGVPQ INEQMLFHGT SSEFVEAICI HNFDWRINGI HGAVFGKGTY FARDAAYSSR FCKDDIKHGN TFQIHGVSLQ QRHLFRTYKS MFLARVLIGD YINGDSKYMR PPSKDGSYVN LYDSCVDDTW NPKIFVVFDA NQIYPEYLID FH (1-331 aa encoded by BC017569)
Mono-ADP-ribosyltransferase that mediates mono-ADP-ribosylation of target proteins (PubMed:25043379, PubMed:25673562). Plays a role in nuclear envelope stability and nuclear remodeling during spermiogenesis. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosyltransferase diphtheria toxin-like 11; ARTD11; PARP-11; poly (ADP-ribose) polymerase family, member 11; Poly [ADP-ribose] polymerase 11; Protein mono-ADP-ribosyltransferase PARP11
Gene Aliases: 5330431N24Rik; AI851877; ARTD11; C12orf6; HIN1L; MIB006; PARP11
UniProt ID: (Human) Q9NR21, (Mouse) Q8CFF0
Entrez Gene ID: (Human) 57097, (Mouse) 101187
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support