Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EALEEALKG PTDGGQSLEE LSSHFYTVIP HNFGHSQPPP INSPELLQAK KDMLLVLADI ELAQALQAVS EQEKTVEEVP HPLDRDYQLL KCQLQLLDSG APEYKVIQTY LEQTGSNHRC PTLQHIWKVN QEGEEDRFQA HSKLGNRKLL WHGTNMAVVA AILTSGLRIM PHSGGRVGKG IYFASENSKS AGYVIGMKCG AHHVGYMFLG EVALGREHHI NTDNPSLKSP PPGFDSVIAR GHTEPDPTQD TELELDGQQV VVPQGQPVPC PEFSSSTFSQ SEYLIYQESQ CRLRYLLEVH L (234-533 aa encoded by BC014260)
PARP3, poly(ADP-ribosyl)transferase 3, belongs to the PARP family. These enzymes modify nuclear proteins by poly-ADP-ribosylation, which is required for DNA repair, regulation of apoptosis, and maintenance of genomic stability. This protein is preferentially localized to the daughter centriole throughout the cell cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A DP-ribosyltransferase (NAD+; poly (ADP-ribose polymerase)-like 3; ADP-ribosyltransferase (NAD+, poly (ADP-ribose polymerase)-like 3; ADP-ribosyltransferase (NAD+; poly (ADP-ribose polymerase)-like 3; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 3; ADP-ribosyltransferase diphtheria toxin-like 3; ADPRT-3; ARTD3; DNA ADP-ribosyltransferase PARP3; IRT1; NAD(+) ADP-ribosyltransferase 3; NAD+ ADP-ribosyltransferase 3; pADPRT-3; PARP-3; poly (ADP-ribose) polymerase family, member 3; poly (ADP-ribosyl) transferase-like 3; Poly [ADP-ribose] polymerase 3; poly(ADP-ribose) synthetase-3; Poly[ADP-ribose] synthase 3; poly[ADP-ribose] synthetase 3; Poly[ADP-ribose] synthetase-3; Protein mono-ADP-ribosyltransferase PARP3
Gene Aliases: A930002C11Rik; ADPRT3; ADPRTL2; ADPRTL3; ARTD3; AW990611; IRT1; PADPRT-3; PARP-3; PARP3
UniProt ID: (Human) Q9Y6F1, (Mouse) Q3ULW8
Entrez Gene ID: (Human) 10039, (Mouse) 235587, (Rat) 300985
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support