Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MKKDESFLGK LGGTLARKRR AREVSDLQEE GKNAINSPMS PALADVHPED TQLEENEERT MIDPTSKEDP KFKELVKVLL DWINDVLVEE RIIVKQLEED LYDGQVLQKL LEKLAGCKLN VAEVTQSEIG QKQKLQTVLE AVHDLLRPRG WALRWSVDSI HGKNLVAILH LLVSLAMHFR APIRLPEHVT VQVVVVRKRE GLLHSSHISE ELTTTTEMMM GRFERDAFDT LFDHAPDKLS VVKKSLITFV NKHLNKLNLE VTELETQFAD GVYLVLLMGL LEDYFVPLHH FYLTPESFDQ KVHNVSFAFE LMLDGGLKKP KARPEDVVNL DLKSTLRVLY NLFTKYKNVE
Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts. Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Affixin; Beta-parvin; parvin, beta
Gene Aliases: CGI-56; PARVB
UniProt ID: (Human) Q9HBI1
Entrez Gene ID: (Human) 29780
Molecular Function:
actin and actin related protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support