Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts. Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Affixin; Beta-parvin; parvin, beta
Gene Aliases: CGI-56; PARVB
UniProt ID: (Human) Q9HBI1
Entrez Gene ID: (Human) 29780
Molecular Function:
actin and actin related protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support