Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to PAX1.
The PAX genes, including PAX1, are a highly conserved family of developmental control genes that encode transcription factors and have been shown to play a role in pattern formation during embryogenesis in vertebrates. See PAX7 (MIM 167410) for a discussion of paired box domain genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HuP48; paired box gene 1; Paired box protein Pax-1; paired domain gene HuP48; wavy tail
Gene Aliases: hbs; hunchback; HUP48; OFC2; Pax-1; PAX1; un; undulated; wt
UniProt ID: (Human) P15863, (Mouse) P09084
Entrez Gene ID: (Human) 5075, (Mouse) 18503
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support