Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 10
PA1-107 has successfully been used in Western blot for detecting overexpressed and recombinant human Pax3 protein, but may not be suitable for detecting endogenous Pax3.
By immunofluorescence, PA1-107 has been used to stain frozen embryonic mouse brain tissue.
PA1-107 has been validated for IP using overexpressed Pax3 protein.
Pax 3 (Paired Box 3) is a transcription factor that is involved in development of the peripheral nervous system, melanocytes, some vascular smooth muscle, and a number of other derivatives. It regulates neurogenesis in pre-migratory neural crest cells from the dorsal neural tube, and in myogenic progenitors in the presomitic mesoderm and the hypaxial somites. Alternative splicing results in isoforms having different C-termini.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HuP2; MGC120381; MGC120382; MGC120383; MGC120384; MGC134778; paired box gene 3; paired box homeotic gene 3; Paired box protein Pax-3; paired domain gene 3; paired domain gene HuP2; PAX3/FKHR fusion
Gene Aliases: CDHS; HUP2; Pax-3; PAX3; Sp; splotch; WS1; WS3
UniProt ID: (Human) P23760, (Mouse) P24610
Entrez Gene ID: (Human) 5077, (Mouse) 18505
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support