Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Additional Information: This antibody is expected to recognize isoforms PAX3 (NP_852122.1), PAX3d (NP_852123.1), PAX3e (NP_852124.1), PAX3g (NP_852126.1), PAX3h (NP_852125.1), and PAX3i (NP_001120838.1).
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Pax 3 (Paired Box 3) is a transcription factor that is involved in development of the peripheral nervous system, melanocytes, some vascular smooth muscle, and a number of other derivatives. It regulates neurogenesis in pre-migratory neural crest cells from the dorsal neural tube, and in myogenic progenitors in the presomitic mesoderm and the hypaxial somites. Alternative splicing results in isoforms having different C-termini.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeodomain protein PAX3; HuP2; MGC120381; MGC120382; MGC120383; MGC120384; MGC134778; paired box gene 3; paired box homeotic gene 3; Paired box protein Pax-3; paired domain gene 3; paired domain gene HuP2; PAX3/FKHR fusion
Gene Aliases: CDHS; HUP2; Pax-3; PAX3; Sp; splotch; WS1; WS3
UniProt ID: (Human) P23760, (Mouse) P24610
Entrez Gene ID: (Human) 5077, (Rat) 114502, (Mouse) 18505
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support